Imprimer
Merino Wool Baselayer Leggings, B181
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B181BKRL B181BKRM B181BKRS B181BKRXL B181BKRXXL
PRTWB181BKRLEA 57.1 EUR
PRTWB181BKRL PRTWB181BKRM PRTWB181BKRS PRTWB181BKRXL PRTWB181BKRXXL
Merino Wool Baselayer Leggings, B181
Tenue de travail
The high content of merino wool contained in this legging offers enhanced thermal protection. The moisture wicking and anti-odour properties keeps the wearer warm, dry and comfortable under exerting conditions.
  • Provides insulation and thermal protection
  • Designed with a comfort fit
  • Mulesling free
  • Flat lock seams for extra comfort
  • Naturally moisture wicking and breathable
  • Lightweight for enhanced wearer comfort
  • Anti-odour
  • Highly durable and long lasting
  • Ideal for all seasons
  • Antimicrobial and anti-odour properties
  • Retail box which aids presentation for retail sales
Baselayer garments are made from a 100% wicking polyester which draws moisture away from the body keeping the wearer cool, dry and comfortable under exerting conditions. The garments are suitable for all levels of activity and can be worn as an extra thermal layer for workwear in cold weather or as a technical sports garment for added comfort and enhanced performance.
All weather protection is engineered with exceptional fabrics and designs, allowing an easy transition between work and leisure. Tested to withstand the rigours of everyday wear, each garment has been designed and manufactured using the best quality materials and high technology construction techniques to ensure total satisfaction with wear after wear.
Height: 220 mm