Imprimer
Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30031.02 SH30015.02 SH30015.03 SH30016.02 SH30030.02 SH30031.01 SH30031.03 SH30031.FS SH30268.02 SH30268.LS SH30588.01 SH30588.02
HYCLSH30031.02EA 29.7 EUR
HYCLSH30031.02 HYCLSH30015.02 HYCLSH30015.03 HYCLSH30016.02 HYCLSH30030.02 HYCLSH30031.01 HYCLSH30031.03 HYCLSH30031.FS HYCLSH30268.02 HYCLSH30268.LS HYCLSH30588.01 HYCLSH30588.02
Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Tampons Tampons de culture de tissus
Balanced salt solutions for cell culture applications.
  • All formulations incorporate a rigorous specification and approval process
  • Powder formats rehydrate to specified quantity
  • Solutions: 0,1 µm sterile filtered
Buffer cell culture growth with HyClone Hank's Balanced Salt Solutions (HBSS), available in lot sizes that facilitate scale-up and manufacturing.