Imprimer
Drosophila containers
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
217101_605 205101_1500
525-0196EA 141 EUR
525-0196 212-3171
Drosophila containers
Flacons Flacons pour drosophiles
PS, crystal-clear.
  • For single use only
  • Temperature resistance –20 to +60 °C
  • Without Ceaprene stopper (must be ordered separately)
Ideally suited for the cultivation of Drosophila melanogaster.