Imprimer
WHEATON® Battery Powered Crimpers and Decappers
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W225808 W225811 W225812 W225813 W225814 W225815 W225820 W225821 W225822
WHEAW225808EA 1720 EUR
WHEAW225808 WHEAW225811 WHEAW225812 WHEAW225813 WHEAW225814 WHEAW225815 WHEAW225820 WHEAW225821 WHEAW225822
WHEATON® Battery Powered Crimpers and Decappers
Pinces à sertir et pinces à dessertir
These battery powered crimpers and decappers fit standard laboratory vials and seals. They come with a long-lasting battery with lithium ion technology, which allows for up to 800 crimps per charge.
  • Powerful motor with a fast cycle
  • Cushioned ergonomic handle reduces hand fatigue
  • Polished crimping jaws provide consistent sealing
  • Can be operated while charging
A warning light indicates when recharging is necessary. The tool can be recharged in one to two hours with the universal power supply included.
The electronically adjustable cycle is accurate and reproducible and can be adjusted with the control pad on top of the unit. The tools are ergonomically designed to reduce strain and arm injury associated with many manual crimping tools.