Imprimer
Non-Contact Dispensers, I.DOT
Fournisseur: DISPENDIX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D16110022091 D16110024910 D16110026000 D16110021796 D16110021802 D16110021808 D16110021814 D16110024973
735-0642EA 128530 EUR
735-0642 735-0643 735-0644 735-0646 735-0647 735-0648 735-0649 735-0650
Non-Contact Dispensers, I.DOT
Automated Liquid Handling Systems
An accessible research appliance enabling the dispensing of nanoliter and microliter scale volumes to 96-, 384-, and 1536-well plates.
  • Use a fraction of the pipette tips required by traditional automation
  • Dispense reagents with a 1 μl dead volume and run more samples for the same reagent cost
  • Dispense 4 nl to 30 μl into any SBS format destination labware with a high degree of precision
  • Dispense with insight into the actual volume dispensed every time with integrated volume verification
  • Software for dispensing a constant volume, dilution, normalization, and pooling should not be complicated
  • Dispense up to 384 source liquids per run at a speed and precision that will continue to surprise with every use
The 'I.DOT' is a liquid handler that anyone can use regardless of automation experience. The instrument transfers volumes as low as 4 nl with unrivalled precision and speed, while dramatically reducing the laboratory’s plastics and reagent consumption.
Whether dispensing enzymes and beads for NGS or qPCR or adding cells, compounds, and buffers for Assay development or high-throughput screening, the 'I.DOT' offers unrivalled precision and ease of use for efficient and reproducible sample preparation.