Imprimer

Chloramphénicol 99

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

Synonyme(s): Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

B20841.14 B20841.22
B20841.14EA 52.4 EUR
B20841.14 B20841.22
Chloramphénicol 99
Chloramphénicol

Purity: 99+% CAS No.: 56-75-7 Molecular Formula: C11H12Cl2N2O5 Formula Weight: 323.14 Physical Form: Crystalline powder Appearance: White to pale cream Melting Point: 149-152° Beilstein Reference: 2225532 Merck Reference: 14,2077 Optical Rotation: +19° (c=6 in ethanol) MDL No.: MFCD00078159


Inhibitor of translation on the 50S subunit at the peptidyltransferase step

Formule: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Poids moléculaire: 323.13 g/mol
Point d'ébullition: 645
Point de fusion: 149…152 °C
Densité: 1.49 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00078159
Numéro CAS: 56-75-7
EINECS: 200-287-4
Indice Merck: 13,02087

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR