Imprimer
BRAND® BIO-CERT® Robotic Tips, PP
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
733146 734148 734150 734152 733166 734168 734170 734172 733646 734650 734652 734654 733666 734670 734672 734674
BRND733146EA 98.9 EUR
BRND733146 BRND734148 BRND734150 BRND734152 BRND733166 BRND734168 BRND734170 BRND734172 BRND733646 BRND734650 BRND734652 BRND734654 BRND733666 BRND734670 BRND734672 BRND734674
BRAND® BIO-CERT® Robotic Tips, PP
Pointes de pipettes
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses with the pietting robot liquid handling station.
  • Raw materials are free from DiHEMDA und Oleamid
  • Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged
  • No parting agents or demolding aids are used
  • BIO-CERT® Liquid handling quality: Free of human DNA, RNase, DNase, pyrogens, PCR-inhibitors and ATP
For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the liquid handling station pipetting robot.