Imprimer
Hei-PLATE Sensor Packages Expert
Fournisseur: Heidolph Instruments GmbH & Co.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
506-31150-00 506-31151-00
HEID506-31150-00EA 1680 EUR
HEID506-31150-00 HEID506-31151-00
Hei-PLATE Sensor Packages Expert
Agitateurs Agitateurs magnétiques
Hei-PLATE sensor Basic and Advanced package Expert.
Basic package includes: Magnetic stirrer Hei-PLATE Mix'n'Heat Expert (145 mm diameter) - Fast and precise heating - Without overshoot - for sample-friendly work, and Pt1000 temperature sensor for precise temperature regulation.
Advanced package includes: Magnetic stirrer Hei-PLATE Mix'n'Heat Expert (145 mm diameter) - Fast and precise heating - Without overshoot - for sample-friendly work, Pt1000 temperature sensor for precise temperature regulation, Pt1000 holder with cable device for securely attaching the temperature sensor and silicone protective cover for a longer lasting protection of your magnetic stirrer.