Imprimer
DURAN® Sealing Rings, for Polished Sperical Joints (VITON)
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1155355 1155356
SCOT1155355EA 61.3 EUR
SCOT1155355 SCOT1155356
DURAN® Sealing Rings, for Polished Sperical Joints (VITON)
Scellés Bagues d'étanchéité
Ball joints without ground joints only seal via the sealing ring.
  • Non autoclavable
  • Used in vacuum applications
Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.