Imprimer
ThermoSafe® Pre-Qualified Envoy® Dry Ice Shippers
Fournisseur: Sonoco Thermosafe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E12F4 E3F4
SCATE12F4EA 74.2 EUR
SCATE12F4 SCATE3F4
ThermoSafe® Pre-Qualified Envoy® Dry Ice Shippers
Boîtes d'expédition et de transport
Envoy® is a range of pre-qualified EPS shippers. Utilising EPS insulation, they are cost-effective and lightweight.
  • Pre-qualified dry ice shipper for deep frozen shipments
  • Core pack included
Envoy® pre-qualified EPS shippers provide excellent thermal protection to slow down the rate of dry ice sublimation. Available in varied sizes, the large version is qualified for 114+ hours at <−20 °C using 18,6 kg of dry ice pellets. The small version is qualified for 104+ hours at <−20 °C using 9,1 kg of dry ice pellets.
Qualification data is available to allow for quick and easy implementation of any project.