Imprimer
Cleanroom ESD Ergo Chairs
Fournisseur: Palbam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PCKD0100 PCKD0110 PCKD0120
PALBPCKD0100EA 1860 EUR
PALBPCKD0100 PALBPCKD0110 PALBPCKD0120
Cleanroom ESD Ergo Chairs
Chaises
Cleanroom ESD ergo chair which lives up to our reputation for high quality and ergonomics/operator comfort.
  • Seat and backrest material and colour: Polyurethane (PU), Black (RAL 9011)
  • Base Material: Chrome steel
Seat height adjustable from 450 to 640 mm.
The chairs include the 'automatic weight adjustment' ergo feature and a tapering backrest offering optimum freedom for shoulders. Options available for conductive glides or casters.
Certifications: The chairs comply with: Cleanliness class 4 DIN EN ISO 14644-1 Anti-static and conductive accordingEn savoir plus