Imprimer
Oil Free Turbomolecular Pumping Systems, CDK
Fournisseur: Welch by Gardner Denver
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
101251 101250
ILMV101251EA 10440 EUR
ILMV101251 ILMV101250
Oil Free Turbomolecular Pumping Systems, CDK
Pompe Pompes à vide à membrane
CDK turbomolecular pumping systems are completely oil-free. It incorporate an oil-free SST turbomolecular pump using dry-running, solid-lubricated ceramic bearings. The backing pump is an oil-free diaphragm vacuum pump.
  • Unique, built-in automatic shut-off device for the backing pump
  • User friendly
  • Automatic shut-off device for the backing pump
  • Lightweight, compact and portable
If the pump system works continually at the ultimate pressure, and if the connected vacuum system has a low real and virtual leak rate (wall degassing), then it is possible that the backing pump will remain switched off for hours.
The CDK systems have all components housed inside a casing, making them very compact and portable. The components include turbomolecular pump, backing pump, controller, and connecting tubing. A simple On/Off button makes operation simple. CDK systems employ three-stage chemical duty diaphragm pump as the backing pump.