Imprimer
Cleanroom Wipes, MicroPure
Fournisseur: SONTARA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1000001885 1000003957 1000003838 1000001898_U 1000003980 1000001913 1000001912
SONT1000001885EA 906 EUR
SONT1000001885 SONT1000003957 SONT1000003838 SONT1000001898_U SONT1000003980 SONT1000001913 SONT1000001912
Cleanroom Wipes, MicroPure
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
Sontara® Cleanroom wipes are specially designed for the challenging task of cleaning a cleanroom - ideal for electronic manufacturing, pharmaceutical, biotechnology applications and surface mount technology. These micro-level cleaning fabrics are 'virtually' lint-free and naturally absorbent, without binders or other additives.
  • Exceptionally clean wipe family with extremely low particle generation, low extractables and ion levels
  • Made with no binders, chemicals or adhesives
  • Special pattern for excellent pick-up
  • Excellent wet and dry strength
  • Solvent resistant
  • Extremely soft hand feel
  • Product traceability
The Sontara® cleanroom products help minimise the risk of contamination that meet all required levels of cleanliness and wiping needs.