Imprimer
ESD Safety Shoes, VD PRO 1500
Fournisseur: Basan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VDPRO1500ES-42 VDPRO1500ES-45 VDPRO1500ES-43 VDPRO1500ES-37 VDPRO1500ES-39 VDPRO1500ES-46
BSNCVDPRO1500ES-42EA 234 EUR
BSNCVDPRO1500ES-42 BSNCVDPRO1500ES-45 BSNCVDPRO1500ES-43 BSNCVDPRO1500ES-37 BSNCVDPRO1500ES-39 BSNCVDPRO1500ES-46
ESD Safety Shoes, VD PRO 1500
Chaussures et bottes
VD PRO 1500 is a light-weight microfibre, seamless shoe, with dust flap, shaft applications and aluminium toe cap, with breathable functional lining, and leather insole.
  • Ultralight microfibre
  • Comfortable fit due to seamless shoe design
  • Dust guard tongue
  • Full-length, removable comfort insole ERGO-SOFT ESD with cushioning system and excellent moisture transportation
Outsole:
TPU Athletic ESD, light blue
Outsole with a profile of 3,5 mm
With increased protection against twisted ankles, reduced lever arms
Supports the natural movement
Light and flexible
Very good scuff resistance and anti-slip properties
Heat-resistant up to approximately 120 °C
VARIO Multiflex® Technology Cushioning:
Lever arms of the shoe force the foot to make unnatural movements and can lead to foot and Achilles tendon complaints, knee pain and back problems. Furthermore, lever arm forces can increase the risk of injuries such as sprained ankles. Lever arm reduced shoes are characterised by lower heel heights, increased insertion depth, softer material and lesser thickness of the sole, which is rounded in the peripheral areas. A result of the VARIO Multiflex® technology is the multiflexible outsole.
Certifications: EN ISO 20345:2011 S2 SRC.