Imprimer
CPC® NS6 Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Fournisseur: CPC COLDER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NS6D17006 NS6D22006 NS6D42006
CPCCNS6D17006EA 259 EUR
CPCCNS6D17006 CPCCNS6D22006 CPCCNS6D42006
CPC® NS6 Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Raccords et connecteurs
NS6 Series couplings feature non-spill valves at a great price. Use the NS6 when even a few drops of spillage pose problems regarding safety, media cost or environmental regulations. These innovative couplings are lightweight, chemically resistant and easy to use.
  • Non-spill design - Disconnect under pressure with no spills
  • Colour coding - Instant visual differentiation of media lines
  • Glass-filled polypropylene - Durable and compatible with many chemicals
  • CPC thumb latch - One-hand connection and disconnection
The non-spill design virtually eliminates spills, minimises downtime and enhances operator safety. Soft touch overmold makes them comfortable in the hand and very attractive.
Material finish: Moulded grey
Mounting option: Thread
Seal options: EPDM
Valve spring: 316 Stainless steel
Valve option: Valved
Spillage: ~0,03 cc per disconnect, 0,42 per connect
Lubricants: Krytox® PFPE (Inert)