Imprimer
Eppendorf LoBind® twin.tec® PCR Plates
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0030129.504 0030129.512 0030129.547 0030129.555 0030129.563 0030129.598 0030129.660 0030129.679
737-0185EA 182 EUR
737-0185 737-0186 EPPE0030129.547 EPPE0030129.555 EPPE0030129.563 731-0705 731-0706 EPPE0030129.679
Eppendorf LoBind® twin.tec® PCR Plates
Microplaques Plaques PCR
Eppendorf twin.tec® PCR plates LoBind, available in 96 and 384-well formats, maximize nucleic acid recovery and are certified as 'PCR clean' for consistently reliable results.
  • LoBind material guarantees maximum sample recovery for improved assay results
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Available in 96- and 384-well formats, twin.tec® PCR plates LoBind® improve recovery of nucleic acids by reducing their adsorption to the wall of the wells. A combination of special manufacturing technologies and selected polypropylene batches ensures that nearly 100 % of DNA and RNA molecules are recovered without the use of surface coatings, thereby eliminating the risk of sample contamination.
Offering maximum confidence in results, all Eppendorf twin.tec PCR plates LoBind are'‘PCR clean'. This means the plates are batch-tested and certified by an independent laboratory to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors.
Autoclavable: 121 °C, 20 min.
Centrifugation stability: 6000×g
Purity grade: PCR clean (nonsterile)