Imprimer
Table Top Refrigerators
Fournisseur: Liebherr
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RE1000 KTE550 KTE551
LIEBRE1000EA 629 EUR
LIEBRE1000 LIEBKTE550 LIEBKTE551
Table Top Refrigerators
Réfrigérateurs
Table-top refrigerators for general use.
  • Very cool
  • Interior LED lighting
  • TouchControl
  • Durable work surface
  • Slim handle equipped with an integrated opening mechanism
  • Dishwasher safe equipment
  • Satin glass shelves with decorative stainless steel strips
These refrigerators comes with screen lock, door alarm, height adjustable door shelves, unclipable door seal, reversible door stop, egg tray and vegetable drawers.