Imprimer
T4 UvsX DNA Recombinase
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-1989 786-1990
GENO786-1989EA 508 EUR
GENO786-1989 GENO786-1990
T4 UvsX DNA Recombinase
Réactifs pour acides nucléiques Nucleic Acid Modification and Labelling
T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes. The protein forms a right-handed nucleoprotein complex on single ssDNA called the presynaptic filament that can search for homology in duplex DNA and pair the recombining DNA molecules to form a DNA joint molecule. This process is aided by ssDNA binding proteins and recombination mediators. The filament is then elongated by DNA polymerase with the newly synthesised strand displacing the old strand. The newly synthesised duplex DNA can then act as a template for the next cycle to achieve exponential amplification of dsDNA. E.coli cells expressing recombinant T4 UvsX gene.
  • UvsX DNA recombinase is free from detectable nuclease contamination
Cdt: Product includes T4 UvsX DNA recombinase 10X UvsX DNA recombinase reaction buffer.