Imprimer
Cell Culture Plates, Nunc™ Multidishes with Nunclon™ Supra Surface
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
140680 140681 140682 140683
734-3644EA 5.1 EUR
734-3644 734-3633 734-3634 734-3645
Cell Culture Plates, Nunc™ Multidishes with Nunclon™ Supra Surface
Microplaques Microplaques pour culture cellulaire
Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
  • Consistent lot-to-lot performance
  • Single-use, flat bottom, with lid
  • Sterile with SAL 10⁻⁶
  • Non-cytotoxic and non-pyrogenic
  • Biocompatible per USP Class VI for raw materials
  • Shelf life: 5 years
The Nunclon Supra surface supports serum-free andEn savoir plus