Imprimer
Erlenmeyer Flasks, Narrow Neck, with Rim
Fournisseur: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
75500 75501 75502 75503
TSLA75500EA 87.4 EUR
TSLA75500 TSLA75501 TSLA75502 TSLA75503
Erlenmeyer Flasks, Narrow Neck, with Rim
Ballons Flacons Erlenmeyers
Temperature-resistant elenmeyer flasks made from transparent quartz glass.
  • Excellent chenical resistance
  • High resistance against thermal shock
  • Made for tasks up to 1050 °C
  • High purity
TQS quartz glass erlenmeyer flasks are made of transparent material for work up to 1050 °C (maximum temperatures up to 1280 °C). High chemical resistance and purity distinguish our quartz glass products. The shelf life of this product is highly dependent on its use and application. With exclusive use up to 1050 °C particularly long service life can be achieved.