Imprimer
Stainless Steel Table with Diamond Rear Tie Bar
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TLTA-CT15X60 TLTARRUPM60X60 TLTARRUP15X60
TEKMTLTA-CT15X60EA 0 EUR
TEKMTLTA-CT15X60 TEKMTLTARRUPM60X60 TEKMTLTARRUP15X60
Stainless Steel Table with Diamond Rear Tie Bar
Mobilier Tables
Teknomek's 304L-grade stainless steel diamond rear tie bar tables combine the best of versatility and contamination resistance. Ideal to place against a wall for added leg room or storage space.
  • Brushed dull polish finish
  • 1,5 mm thick stainless steel sheet
  • 4× zinc thread with plastic floor levelling adjustable feet (Static)
  • 4× stainless steel castors (two braked) for mobile
  • 100 kg payload
The stainless steel table with diamond rear tie bar are a very versatile work surface and storage solution for industries that require sterile, hygienic and contamination resistant surfaces. The table has been intelligently designed to be easy to clean, reducing harbouring surfaces and removing potential trap points.
Easy clean features include diamond rear tie bar which has been raised to 300 mm from the ground to aid ease of cleaning and also to allow for extra space underneath so it can be utilised for extra storage. The rear legs are inset 100 mm to allow clearance from skirting boards or where floors have returned up walls.
Finished in brushed dull polish, these tables come with 304L-grade stainless steel construction with an upstand including plastic floor levelling adjustable feet, or stainless steel castors. The sturdy 30 mm box frame grants robustness to the table so you don't have to worry about payloads.