Imprimer
Stainless Steel Hygienox Table with Tubular Rear Tie Bar
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TLTAHYRRM12X60 TLTAHYEPR9060 TLTAHYEPR1260 TLTAHYEPR1560
TEKMTLTAHYRRM12X60EA 0 EUR
TEKMTLTAHYRRM12X60 TEKMTLTAHYEPR9060 TEKMTLTAHYEPR1260 TEKMTLTAHYEPR1560
Stainless Steel Hygienox Table with Tubular Rear Tie Bar
Mobilier Tables
Hygienox 316-grade stainless steel tables are made specifically for cleanroom environments, giving extreme protection against pitting and crevice corrosion caused by harsh clean down routines.
  • Fully welded 316-grade stainless steel
  • 316-grade stainless steel feet, with non-marking anti-slip rubber base and no exposed thread
  • Chemically and physically resistant
  • Additional cleaning, pickling and polishing over above normal Teknomek standards (All visible and non visible welds pickled and passivated)
  • 100 kg payload
Providing protection from corrosiveEn savoir plus