Imprimer
Cerex® C18 SPE Heavy Load Columns
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
602-1003 618-0201R
TCAT602-1003EA 0 EUR
TCAT602-1003 TCAT618-0201R
Cerex® C18 SPE Heavy Load Columns
Colonnes pour chromatographie
Endcapped C18 chemistry.
  • Reversed-phase
  • Liquid handler compatible
A 10 µm micro particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation.