Imprimer
Resolvex® Column Temperature Controller
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
282-0240 282-0252 282-0300
TCAT282-0240EA 0 EUR
TCAT282-0240 TCAT282-0252 TCAT282-0300
Resolvex® Column Temperature Controller
Appareils chauffants Fours colonne
Column temperature controllers for heating and cooling.
  • Reliable PID microprocessor controlled thermal circuit stabilizes the operating environment
  • Easily-seated thermal blanket encloses the column in horizontal, vertical or slant orientation
  • Heater inserts are modular for easy removal and arrangement
Based on a unique, space-saving concept for efficient temperature control of liquid chromatography columns. Column enclosure accepts ³/₈" ext. Ø HPLC columns of any length up to 30 centimeters (15 centimeters in Column Heater 150).