Imprimer
Cubis® II MCA 225S Ultra-High Resolution Semi-Micro Balances
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Cubis® II
SARBMCA225S3CF34HEA 0 EUR
SARBMCA225S3CF34H SARBMCA225S3CF34I SARBMCA225S3CE13I SARBMCA225S3CE13H SARBMCA225S3CE4 SARBMCA225S3CE3 SARBMCA225S3CE2 SARBMCA225S3CF1M SARBMCA225S3CF1I SARBMCA225S3CEI SARBMCA225S3CF24 SARBMCA225S3CF23 SARBMCA225S3CE12M SARBMCA225S3CE24M SARBMCA225S3CE14H SARBMCA225S3CF23H SARBMCA225S3CF23I SARBMCA225S3CF99I SARBMCA225S3CE14I SARBMCA225S3CE124H SARBMCA225S3CF2M SARBMCA225S3CF2H SARBMCA225S3CF234 SARBMCA225S3CF2I SARBMCA225S3CE14 SARBMCA225S3CE123H SARBMCA225S3CF34 SARBMCA225S3CE123I SARBMCA225S3CE13 SARBMCA225S3CE13M SARBMCA225S3CF34M SARBMCA225S3CE123M SARBMCA225S3CE12 SARBMCA225S3S0123I SARBMCA225S3CE99I SARBMCA225S3CFM SARBMCA225S3S0123H SARBMCA225S3CE23I SARBMCA225S3CE23H SARBMCA225S3CE234H SARBMCA225S3S023I SARBMCA225S3S023H SARBMCA225S3CE234 SARBMCA225S3S023M SARBMCA225S3S099I SARBMCA225S3CE124I SARBMCA225S3CE34M SARBMCA225S3S099M SARBMCA225S3CE124M SARBMCA225S3S0124 SARBMCA225S3S0123 SARBMCA225S3CF2 SARBMCA225S3CEM SARBMCA225S3CE12I SARBMCA225S3CE24I SARBMCA225S3CE12H SARBMCA225S3CE24H SARBMCA225S3CF4 SARBMCA225S3CF3 SARBMCA225S3S034I SARBMCA225S3S034H SARBMCA225S3CE124 SARBMCA225S3CE123 SARBMCA225S3S034M SARBMCA225S3CE99M SARBMCA225S3CF13 SARBMCA225S3CFI SARBMCA225S3CF12 SARBMCA225S3CE234I SARBMCA225S3CE23M SARBMCA225S3CF14 SARBMCA225S3S0234 SARBMCA225S3CE234M SARBMCA225S3CF123M SARBMCA225S3S0234M SARBMCA225S3CF14M SARBMCA225S3S03M SARBMCA225S3S0234H SARBMCA225S3CF123I SARBMCA225S3S0234I SARBMCA225S3S02 SARBMCA225S3S03 SARBMCA225S3CF14H SARBMCA225S3S03H SARBMCA225S3S03I SARBMCA225S3S04 SARBMCA225S3CF123H SARBMCA225S3CF14I SARBMCA225S3CE4I SARBMCA225S3CE4H SARBMCA225S3S013I SARBMCA225S3CE3M SARBMCA225S3S013H SARBMCA225S3S013M SARBMCA225S3CE3I SARBMCA225S3S034 SARBMCA225S3S02M SARBMCA225S3CF13M SARBMCA225S3S0124M SARBMCA225S3S04M SARBMCA225S3S0124H SARBMCA225S3S0124I SARBMCA225S3CE34I SARBMCA225S3S04H SARBMCA225S3S04I SARBMCA225S3CE34H SARBMCA225S3S012I SARBMCA225S3S012H SARBMCA225S3S024I SARBMCA225S3S024H SARBMCA225S3S012M SARBMCA225S3S024M SARBMCA225S3CE4M SARBMCA225S3S0123M SARBMCA225S3CF24I SARBMCA225S3CF12H SARBMCA225S3CF12I SARBMCA225S3S01I SARBMCA225S3CF234M SARBMCA225S3CF234I SARBMCA225S3CF234H SARBMCA225S3CF24H SARBMCA225S3CF4H SARBMCA225S3CF3M SARBMCA225S3CE1M SARBMCA225S3CF3I SARBMCA225S3S014 SARBMCA225S3CF124 SARBMCA225S3CF3H SARBMCA225S3CE1I SARBMCA225S3CE24 SARBMCA225S3S012 SARBMCA225S3S013 SARBMCA225S3CF123 SARBMCA225S3CE14M SARBMCA225S3CF23M SARBMCA225S3CE23 SARBMCA225S3CF99M SARBMCA225S3S0M SARBMCA225S3CF13I SARBMCA225S3S02I SARBMCA225S3CF124M SARBMCA225S3CF124H SARBMCA225S3S02H SARBMCA225S3CF13H SARBMCA225S3CF124I SARBMCA225S3CE3H SARBMCA225S3S0I SARBMCA225S3CE2M SARBMCA225S3CF4M SARBMCA225S3S014I SARBMCA225S3S014H SARBMCA225S3CE2I SARBMCA225S3CE2H SARBMCA225S3CF4I SARBMCA225S3S014M SARBMCA225S3S023 SARBMCA225S3S024 SARBMCA225S3CF24M SARBMCA225S3S01M SARBMCA225S3CF12M SARBMCA225S3CE34
Cubis® II MCA 225S Ultra-High Resolution Semi-Micro Balances
Balances
The Cubis® II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions.
  • Draft shield (D) for high capacity micro balances with upgradable hardware
  • Superior weighing performance
  • No more issues with charged samples
  • Cleaning process guidance
  • Hardware upgradeability, compliance and data integrity
  • Easy to use with motorised inner draft shield
MCA display - 7" colour touch TFT display in 16:9 format with intuitive user interface. Factory installed basic set of weighing applications (license free) and licensable software packages (QPs) for various applications (QApps) and functional extensions.
Draft shield (D) - Manual glass analytical draft shield chamber, with smooth-action doors that open wide and provide unimpeded access to the weighing chamber.
Hardware options: All hardware licenses (HWL), Ioniser (ION) and Motorised draft shield (MDS).
Intuitive process for daily and advanced cleaning is now available in all Cubis® II MCA. The cleaning QApp provides visual guidance, information about chemical compatibility as well as electronical tracking of these events. Hardware features like motorised draft shield function or built-in ioniser can be activated post-purchase. An automated inner draft shield is available as a klick-in accessory which is recommended to use under challenging environmental conditions (e.g. in drafty environment).
Installation of Cubis® II balances in a workbench or laminar hood with filtered air flow subjects the instrument to drafty conditions. The motorised inner draft shield ensures best weighing performances and ease of use even under drafty conditions.
Fast measurement time and accurate weighing results are guaranteed thanks to the next generation monolithic weighing system and the engineering solution compensating for environmental changes. The glass draft shields coated with conductive layer prevents from outer electrostatic effects and the built-in ioniser effectively eliminates electrostatic charges from samples.
The end-to-end data integrity, technical controls for 21 CFR Part 11 compliance, integrated audit-trail and state-of-the-art user management ensure laboratories can meet regulatory requirements. These features are directly available on the Cubis® II balances, without the need of additional software. The Ingenix Suite is a flexible, open solution that works with or without an ELN/LIM system. It offers unlimited connections to easily manage the entire Cubis® II MCA lab balance fleet across all labs within the same network.
The teaching function with learning capability of the motorised draft shield, guided workflows for many different applications (QApps), automated motorised levelling and automatic internal adjustment (isoCAL) help to with easy and error-free operation of the balance.
QApp Pharma software application package (QP1): It contains applications concerning the topic compliance with pharmaceutical-relevant guidelines as 21 CFR Part 11 and USP 39, Chapter 41. The Pharma package includes applications such as user management, digital signatures, audit trail, USP minimum weight.
QApp Advanced software application package (QP2): It includes various complex weighing applications includes evaluation. This includes applications used for density determination, percentage weighing, counting, back weighing, residual dirt analysis, residue on ignition, sieve analysis, filter weighing, check weighing, formulation, averaging, and so on.
QApp Utility software application package (QP3): It contains weighing applications and function extensions such as bootscreen, colour scheme, free formula, fibre coarseness, diameter determination, air buoyancy correction, paper weight, statistics and printing of QR|bar codes.
QApp Connectivity software application package (QP4): It includes applications for data exchange, for example to Windows file server, FTPS, StarLims, and so on.
QApp software application package all inclusive (QP99): It includes four different sub-packages for compliance (pharma (QP1)), complex weighing applications (advanced (QP2)), weighing applications and helpful tools (utilities (QP3)) and connectors for data exchange (connectivity (QP4)).