Imprimer
Cubis® II MCA 225P Ultra-High Resolution Semi-Micro Balances
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Cubis® II
SARBMCA225P3S034IEA 19150 EUR
SARBMCA225P3S034I SARBMCA225P3S034M SARBMCA225P3S0234 SARBMCA225P3CF234 SARBMCA225P3CE14H SARBMCA225P3CF23H SARBMCA225P3CE14I SARBMCA225P3CF99I SARBMCA225P3CF23I SARBMCA225P3CE14M SARBMCA225P3CEUDMD SARBMCA225P3CF99M SARBMCA225P3CF23M SARBMCA225P3S04M SARBMCA225P3S023H SARBMCA225P3S023I SARBMCA225P3S04I SARBMCA225P3S099I SARBMCA225P3S04H SARBMCA225P3CFM SARBMCA225P3S023M SARBMCA225P3S099M SARBMCA225P3CFI SARBMCA225P3S0123 SARBMCA225P3S0124 SARBMCA225P3CF34H SARBMCA225P3CE4M SARBMCA225P3CE13H SARBMCA225P3CE13I SARBMCA225P3CF34I SARBMCA225P3CE4I SARBMCA225P3CE13M SARBMCA225P3CE4H SARBMCA225P3CF34M SARBMCA225P3CF3 SARBMCA225P3CF2 SARBMCA225P3CF4 SARBMCA225P3CEM SARBMCA225P3S024H SARBMCA225P3S024I SARBMCA225P3S012H SARBMCA225P3S03M 611-5807 SARBMCA225P3S012I SARBMCA225P3CEI 611-5806 611-5809 SARBMCA225P3S024M 611-5808 SARBMCA225P3S0123H SARBMCA225P3S012M SARBMCA225P3S0123I SARBMCA225P3CF24 SARBMCA225P3CF13I SARBMCA225P3CF1I SARBMCA225P3CF13H SARBMCA225P3CF13M 611-5773 611-5772 611-5775 611-5774 SARBMCA225P3CF23 611-5810 SARBMCA225P3CE2 611-5776 611-5812 SARBMCA225P3CE4 SARBMCA225P3CE3 611-5811 611-5814 611-5813 SARBMCA225P3CF1M 611-5816 SARBMCA225P3S013H 611-5815 SARBMCA225P3S013I 611-5817 SARBMCA225P3S013M SARBMCA225P3S0124I SARBMCA225P3CF14 SARBMCA225P3CF13 SARBMCA225P3S0124H SARBMCA225P3CF124 SARBMCA225P3CF123 SARBMCA225P3CF12H SARBMCA225P3CF24I SARBMCA225P3CF24H SARBMCA225P3CF12I SARBMCA225P3CF24M SARBMCA225P3CF12M SARBMCA225P3CF12 SARBMCA225P3S0123M SARBMCA225P3S014H SARBMCA225P3S014I SARBMCA225P3S014M SARBMCA225P3S0234I SARBMCA225P3S0234H SARBMCA225P3CF123I SARBMCA225P3CF123H SARBMCA225P3CF123M SARBMCA225P3CEUDHW SARBMCA225P3CE34H SARBMCA225P3CF3I SARBMCA225P3CE34I SARBMCA225P3CE1I SARBMCA225P3CF3H SARBMCA225P3S012 SARBMCA225P3S014 SARBMCA225P3CE34M SARBMCA225P3S013 SARBMCA225P3S0124M SARBMCA225P3CE23 SARBMCA225P3CE24 SARBMCA225P3CE1M SARBMCA225P3CF3M SARBMCA225P3CF124H SARBMCA225P3CE14 SARBMCA225P3CF124I SARBMCA225P3CF124M SARBMCA225P3CF14I SARBMCA225P3CF2I SARBMCA225P3CE234M SARBMCA225P3CF2H SARBMCA225P3CF14M SARBMCA225P3CE234I SARBMCA225P3CE234H SARBMCA225P3CF34 SARBMCA225P3CE12 SARBMCA225P3S0M SARBMCA225P3S0234M SARBMCA225P3CE13 SARBMCA225P3S0I 611-5805 SARBMCA225P3CF14H SARBMCA225P3CF2M SARBMCA225P3S03H SARBMCA225P3CF234H SARBMCA225P3S03I SARBMCA225P3CF234I SARBMCA225P3CE99M SARBMCA225P3CF234M SARBMCA225P3CE123 SARBMCA225P3CE124 SARBMCA225P3S04 SARBMCA225P3CE3M SARBMCA225P3CE24H SARBMCA225P3CE12H SARBMCA225P3CE24I SARBMCA225P3CE124M SARBMCA225P3CE12I SARBMCA225P3S034 SARBMCA225P3CE3H SARBMCA225P3CE3I SARBMCA225P3S03 SARBMCA225P3CE124I SARBMCA225P3CE24M SARBMCA225P3CE12M SARBMCA225P3S02 SARBMCA225P3CE124H SARBMCA225P3S02M SARBMCA225P3S02I SARBMCA225P3S02H SARBMCA225P3CE234 SARBMCA225P3CF4M SARBMCA225P3CE123I SARBMCA225P3CE23H SARBMCA225P3CE2I SARBMCA225P3CE23I SARBMCA225P3CF4I SARBMCA225P3S023 SARBMCA225P3CE99I SARBMCA225P3CF4H SARBMCA225P3CE2H SARBMCA225P3CE123H SARBMCA225P3CE23M SARBMCA225P3S024 SARBMCA225P3CE34 SARBMCA225P3S01M SARBMCA225P3CE123M SARBMCA225P3S01I SARBMCA225P3CE2M SARBMCA225P3S034H
Cubis® II MCA 225P Ultra-High Resolution Semi-Micro Balances
Balances
The Cubis® II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions.
  • Draft shield (D) for high capacity micro balances with upgradable hardware
  • Superior weighing performance
  • No more issues with charged samples
  • Cleaning process guidance
  • Hardware upgradeability, compliance and data integrity
  • Easy to use with motorised inner draft shield
MCA display - 7" colour touch TFT display in 16:9 format with intuitive user interface. Factory installed basic set of weighing applications (license free) and licensable software packages (QPs) for various applications (QApps) and functional extensions.
Draft shield (D) - Manual glass analytical draft shield chamber, with smooth-action doors that open wide and provide unimpeded access to the weighing chamber.
Hardware options: All hardware licenses (HWL), Ioniser (ION) and Motorised draft shield (MDS).
Intuitive process for daily and advanced cleaning is now available in all Cubis® II MCA. The cleaning QApp provides visual guidance, information about chemical compatibility as well as electronical tracking of these events. Hardware features like motorised draft shield function or built-in ioniser can be activated post-purchase. An automated inner draft shield is available as a klick-in accessory which is recommended to use under challenging environmental conditions (e.g. in drafty environment).
Installation of Cubis® II balances in a workbench or laminar hood with filtered air flow subjects the instrument to drafty conditions. The motorised inner draft shield ensures best weighing performances and ease of use even under drafty conditions.
Fast measurement time and accurate weighing results are guaranteed thanks to the next generation monolithic weighing system and the engineering solution compensating for environmental changes. The glass draft shields coated with conductive layer prevents from outer electrostatic effects and the built-in ioniser effectively eliminates electrostatic charges from samples.
The end-to-end data integrity, technical controls for 21 CFR Part 11 compliance, integrated audit-trail and state-of-the-art user management ensure laboratories can meet regulatory requirements. These features are directly available on the Cubis® II balances, without the need of additional software. The Ingenix Suite is a flexible, open solution that works with or without an ELN/LIM system. It offers unlimited connections to easily manage the entire Cubis® II MCA lab balance fleet across all labs within the same network.
The teaching function with learning capability of the motorised draft shield, guided workflows for many different applications (QApps), automated motorised leveling and automatic internal adjustment (isoCAL) help to with easy and error-free operation of the balance.
QApp Pharma software application package (QP1): It contains applications concerning the topic compliance with pharmaceutical-relevant guidelines as 21 CFR Part 11 and USP 39, Chapter 41. The Pharma package includes applications such as user management, digital signatures, audit trail, USP minimum weight.
QApp Advanced software application package (QP2): It includes various complex weighing applications includes evaluation. This includes applications used for density determination, percentage weighing, counting, backweighing, residual dirt analysis, residue on ignition, sieve analysis, filter weighing, checkweighing, formulation, averaging, and so on.
QApp Utility software application package (QP3): It contains weighing applications and function extensions such as bootscreen, color scheme, free formula, fibre coarseness, diameter determination, air buyoancy correction, paper weight, statistics and printing of QR|bar codes.
QApp Connectivity software application package (QP4): It includes applications for data exchange, for example to Windows file server, FTPS, StarLims, and so on.
QApp software application package all inclusive (QP99): It includes four different sub-packages for compliance (pharma (QP1)), complex weighing applications (advanced (QP2)), weighing applications and helpful tools (utilities (QP3)) and connectors for data exchange (connectivity (QP4)).