Imprimer
Crystallising Dishes, without Spout
Fournisseur: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
75170 75171 75172 75173 75174 75175
TSLA75170EA 39.4 EUR
TSLA75170 TSLA75171 TSLA75172 TSLA75173 TSLA75174 TSLA75175
Crystallising Dishes, without Spout
Plateaux/Récipients/Pots Cristallisoirs
Temperature-resistant crystallising dishes made from transparent quartz glass.
  • Excellent chemical resistance
  • High resistance against thermal shock
  • Made for tasks up to 1050 °C
  • High purity
The TQS quartz glass crystallising dishes are made for tasks up to 1050 °C (maximum limit 1280 °C). High chemical resistance and purity characterise these products. Quartz glass products have a long life for frequent use with temperatures up to 1050 °C, but the durability is dependent upon use and applications.