Imprimer
Rotors for Centrifuges, 5810/5810 R
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5810709.008 5810711.002 5810718.007 5810725.003 5810743.001 5820710.004 5820740.000 5820754.001 5820755.008
521-0078EA 2210 EUR
521-0078 522-2730 521-0081 521-0145 522-0036 521-2072 521-1958 521-2074 521-2075
Rotors for Centrifuges, 5810/5810 R
Centrifugeuses Rotors
Eppendorf Rotors for centrifuges 5810/5810 R - precision redefined. Tailor your high-capacity centrifugation with varied rotor options. Choose from a range of capacities and speeds, accessorize with versatile buckets, and ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Redesign your high-capacity centrifugation with Eppendorf rotors crafted for centrifuges 5810/5810 R. These rotors offer a spectrum of capacities and speeds, providing adaptability for various sample processing needs. Accessorize with versatile buckets, ensuring efficient customization of your centrifuge setup. Ideal for diverse applications, the Centrifuge 5810 series is further enhanced with these precision-engineered rotors, guaranteeing accuracy and reproducibility in every spin. Explore the range to optimize your centrifugation experience.