Imprimer
Combitips® advanced Forensic DNA Grade - Individually Blister-Wrapped Positive Displacement Pipette Tips
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0030089.855 0030089.863 0030089.871
613-5797EA 154 EUR
613-5797 613-5798 613-5799
Combitips® advanced Forensic DNA Grade - Individually Blister-Wrapped Positive Displacement Pipette Tips
Pointes de pipettes
Eppendorf Combitips® advanced in 'Forensic DNA Grade' offer unmatched precision and safety, individually blister-wrapped for maximal DNA contamination prevention. Ideal for forensic applications requiring the utmost accuracy and sterility.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0,1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nl
  • Combitips® advanced in ‘Forensic DNA Grade’ quality conform to the requirements of ISO 18385
  • Particularly well suited for forensic applications and the isolation and analysis of DNA
  • Elongated Combitips® advanced in volumes 2,5, 5,0, and 10 ml will reach the bottom of most common laboratory vessels
  • Combitips® advanced are offered in five different purity grades to support a range of applications
  • Appropriate purity grade for each application: Eppendorf Quality, PCR clean, Forensic DNA Grade, sterile, or Biopur®
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The tip of choice when working with Repeater®/Multipette® multi-dispenser pipettes from Eppendorf:
Thanks to their revolutionary design, Combitips® advanced dispenser tips provide many benefits for work in a modern laboratory. With an integrated piston, they function according to the positive displacement principle and ensure that the correct volume of liquid is dispensed regardless of the physical properties of the liquid (e.g., viscosity, volatility, density, temperature). Achieve high-precision dispensing, independent of the flow characteristics of the liquid, and safely work with radioactive or toxic materials due to the hermetically sealed piston that prevents aerosol contamination.
Eppendorf pioneered the Repeater®/Combitips® multi-dispenser system in 1978 and improved it several times since. It was the very first pipetting system allowing users to fill the tip once and dispense multiple times. The concept remains a driver for efficient liquid handling and a staple in many labs working with plates or series of tubes. In 2023, the Combitips® advanced pipette tip line-up has been certified with ACT® Environmental Impact Factor Labels by the My Green Lab® non-profit organisation, making it the ideal choice for labs seeking more sustainable products.
Certifications: ACT® Label and lot-specific certificates for Eppendorf consumables in 'Forensic DNA Grade' quality can be downloaded from the Eppendorf website.
Informations de commande: Combitips® advanced are designed for use with Eppendorf Repeater®/Multipette® multi-dispensers only.