Imprimer
ep Dualfilter T.I.P.S.® Filter Pipette Tips, BioBased Reloads
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0030081.030 0030081.048 0030081.056 0030081.064 0030081.072 0030081.080 0030081.099 0030081.102
613-7262EA 195 EUR
613-7262 613-7263 613-7264 613-7265 613-7266 613-7267 613-7268 613-7269
ep Dualfilter T.I.P.S.® Filter Pipette Tips, BioBased Reloads
Pointes de pipettes
Eppendorf ep Dualfilter T.I.P.S.® BioBased Reloads offer eco-conscious pipetting with tips made from 90% plant-based material, 'PCR clean' and 'Sterile' grades with dual-layer filtration, ensuring unparalleled precision and contamination prevention in sustainable, reloadable formats.
  • Improve the laboratory carbon footprint with ep Dualfilter T.I.P.S.® BioBased in the new Eppendorf Reloads
  • Produced from recycled feedstock in second generation, derived solely from waste and residue streams from vegetable oil production
  • Significantly reduce laboratory waste volume by using Reloads instead of disposable racks
  • Pipette tips with ACT (Accountability, Consistency, and Transparency) certification
  • Premium material, quality, and purity certified
  • Excellent sample integrity as no slip agents, plasticizers, and biocides are used
  • General quality certificates in compliance with standards and general Eppendorf quality items
  • Available in volumes from 10 to 1,250 µl Long in Reloads for use with epT.I.P.S.® Box 2.0
  • Also available as epT.I.P.S.® BioBased Biopur® and ep Dualfilter T.I.P.S.® SealMax® BioBased Biopur®
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
"The Eppendorf epT.I.P.S.® BioBased in volumes up to 1,250µl open up new doors to make laboratory work significantly more sustainable without jeopardising experimental results. By using a second-generation bio-based polymer made from at least 90% renewable resources (e.g. waste and residues from vegetable oil refining or waste cooking oil), the product-related carbon footprint of these pipette tips and filter pipette tips was significantly reduced. "General Quality Certificates" on compliance with standards and general quality requirements, product-specific and lot-specific purity certificates, as well as "ACT Certificates" (the independent environmental impact factor certification by My Green Lab) attest the well-known premium product quality expected from Eppendorf pipette tips, combined with a sustainability factor that will clearly be visible in the laboratory carbon footprint with continuous use of these pipette and filter pipette tips.
Together with the use of more sustainable starting material for the pipette tips, these bio-based tips come in a Reload variant which meets the same purity and sterility requirements as conventional disposable racks. This Reload variant saves up to 54% fossil-based plastic compared to conventional racks and significantly reduces the volume of laboratory waste. The new Reloads fit perfectly into the well-known epT.I.P.S.® Box 2.0.
Thus both aspects – bio-based raw material for the pipette tips and significantly reduced use of plastic for the Reloads – contribute to an improved laboratory carbon footprint and to laboratory waste reduction.
Certifications: Lot-specific certificates for the purity grade PCR clean/Sterile and sustainability declarations according to ISCC PLUS can be downloaded from the Eppendorf website.