Imprimer
ESD Safety Shoes, Low, LUAN, S1PS
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
729121/41 729121/42 729121/43 729121/44 729121/45 729121/46 729121/47 729121/48 729121/39 729121/35 729121/38 729121/36 729121/37 729121/40
ELTN729121/41EA 99.9 EUR
ELTN729121/41 ELTN729121/42 ELTN729121/43 ELTN729121/44 ELTN729121/45 ELTN729121/46 ELTN729121/47 ELTN729121/48 ELTN729121/39 ELTN729121/35 ELTN729121/38 ELTN729121/36 ELTN729121/37 ELTN729121/40
ESD Safety Shoes, Low, LUAN, S1PS
Chaussures et bottes
Mesh material with recycled material content/Microfibre with recycled material content.
  • Breathable lining with recycled material content
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD grey with recycled material content
  • Non-metallic, penetration-resistant midsolewith recycled material content
  • TPU/PU sole MAXXIMO
  • Steel toe cap
Safety shoes are designed for use in areas where there is a risk of electrostatic discharge (ESDS/ESD) and workplaces on hard undergrounds. The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides, creating more energy in every step.
Certifications: EN ISO 20345 S1PS FO/SR, form A.