Imprimer
pH Sensor, Non-glass, Memosens CPS97E
Fournisseur: Endress+Hauser
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CPS97E-AA8BHH2 CPS97E-AA8BHH9
ENDRCPS97E-AA8BHH2EA 982 EUR
ENDRCPS97E-AA8BHH2 ENDRCPS97E-AA8BHH9
pH Sensor, Non-glass, Memosens CPS97E
Électrodes Électrodes pH
Memosens 2.0 ISFET pH sensor for polluted media in chemical processes, paper or pigment production.
  • Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
  • Unbreakable: PEEK sensor body withstands harsh conditions ensuring product safety
  • Open aperture prevents blocking of the sensor even in media with a high dirt load
  • Chemically stable reference gel assures long-term stable measurements
  • Low maintenance: ISFET technology is insensitive to high temperature or temperature changes which results in long calibration intervals
  • Maximum process safety: Non-contact, inductive signal transmission eliminates all problems caused by moisture or corrosion
Memosens CPS97E is the expert for media with a high content of fibres or suspended solids such as dispersions, precipitations and emulsions. Its unbreakable shaft and the open aperture guarantee reliable measurement under harsh process conditions. CPS97E features Memosens 2.0 digital technology, offering extended storage of calibration and process data for predictive maintenance. The sensor resists moisture and enables lab calibration, enhancing process integrity and increasing process uptime.
Application:
Chemical processes
Contaminated media and suspended solids
- Dispersions
- Precipitation
- Emulsions
Pulp and paper industry
Sensor shaft: PEEK
Sealings: FFKM
Metal lead: Ag/AgCl
O-ring: FKM
Process coupling: PPS fiber-glass reinforced
Nameplate: Ceramic metal oxide
Ingress protection: IP 68
Process pressure: Maximum 11 bar abs. at 100 °C
Certifications: With the following approvals for use in potentially explosive areas of Zone 0, Zone 1 and Zone 2: ATEX, IECEx, CSA C / US, NEPSI, JPN Ex, INMETRO, UKCA and Korea Ex.