Imprimer
Fall Arrest Roofing Kits
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PRTWFP66REREA 181 EUR
PRTWFP66RER PRTWFP99BKR
Fall Arrest Roofing Kits
Protections anti-chutes Cordes de sécurité
Kit includes two-point harness, carabiner, 12 mm detachable rope grab, 10 m kernmantle static rope, and nylon drawstring bag.
  • Chest loops
  • High-quality 12 mm kernmantle rope
  • Maximum rated load: 100 kg
The versatile drawstring bag is high-strength yet lightweight, making it ideal for securely holding fall protection products.
10-year shelf life from manufacturing date outlined on product label.