Imprimer
Drums, Stainless Steel, with Toggle Clamps
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A624-1 A624-2 A624-3 A624-5 A624-6 A624-8 A624-10 A624-12 A624-15
SAMPA624-1EA 1130 EUR
SAMPA624-1 SAMPA624-2 SAMPA624-3 SAMPA624-5 SAMPA624-6 SAMPA624-8 SAMPA624-10 SAMPA624-12 SAMPA624-15
Drums, Stainless Steel, with Toggle Clamps
Barils, tonneaux, fûts
A multipurpose stainless steel container available in a range of sizes from 1 to 15 L. Ideal for work in a range of different environments include small scale production, laboratories and general day to day use. They are manufactured from 304 stainless steel and finished to a high quality finish.
  • Multi-purpose drum
  • 304 stainless steel body and lid
  • Swing carrying handle
  • Crevice free interior
They are ideal for working with both powders and liquids. The containers come with a drop-on lid. The lid is secured with toggle clamps and an O-ring seal. Simply pull down the toggle clamps to secure the lid.