Imprimer
QlickSMART SNAPIT Lite Personal Regular Ampoule Openers
Fournisseur: SWANN MORTON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SN-02R SN-02RB SN-02RPK SN-02RPP
SWANSN-02REA 46.1 EUR
SWANSN-02R SWANSN-02RB SWANSN-02RPK SWANSN-02RPP
QlickSMART SNAPIT Lite Personal Regular Ampoule Openers
Ouvre-ampoules
Qlicksmart SnapIT is a simply safer way to open glass ampoules and avoid painful and dangerous sharps injuries. This easy-to-use multi-use ampoule opener avoids the unacceptable sharps injury rates seen when ampoules are opened by hand. L×W: 80×13 mm, 11 g. Plastic (body), Silicon (O-ring), Stainless steel (keyring and spring).
  • With SnapIT you can safely:
  • Insert the ampoule lid into your multi-use SnapIT
  • Snap open the ampoule
  • Eject the ampoule lid directly into a sharps container
  • Not autoclavable
There are both personal and trolley versions available in the range:
Trolley edition: for team use featuring a flat side so it can lay on a bench top or trolley without rolling away - perfect for hospital medication preparation areas.
Pocket edition: anodised aluminium for personal use on a key ring, hung from a lanyard or kept in your pocket. Lite edition: made from sturdy, low cost plastic.
The regular size Snap-It can be used with ampoules of 1 to 2 ml, 5 to 10 ml and 10 to 15 ml whilst the large trolley edition can be used with ampoules of 5 to 10 ml and 20 to 25 ml.
SnapIT should be used wherever glass ampoules are being used: Hospitals, nurses, paramedics, anaesthitists, educational facilities, veterinary practice, dentist surgeries,general practices, home users or anyone that opens
glass ampoules.
SnapIT avoids ampoule sharps injuries by keeping both hands away from the sharp edges during ampoule opening. One hand holds the base of the ampoule while the other hand simply uses the SnapIT to snap off the ampoule lid. The ampoule lid is contained safely within the SnapIT until ejected. Multi-use. O-ring can be replaced if lost or broken.
Certifications: USA FDA listed.