Imprimer

2,4,5-Triméthyloxazole ≥97%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): 2,4,5-Triméthyl-1,3-oxazole

A10328.06 A10328.14
A10328.06EA 48.2 EUR
A10328.06 A10328.14
2,4,5-Triméthyloxazole ≥97%
2,4,5-Triméthyloxazole

Purity: 97% CAS No.: 20662-84-4 Molecular Formula: C6H9NO Formula Weight: 111.14 Storage Temperature: 2°C to 8°C Flash Point: 33°(91°F) Refractive Index: 1.4420 Odour: Pungent Boiling Point: 133-134° Beilstein Reference: 107043 UN No.: 1993 MDL No.: MFCD00005308

Formule: C₆H₉NO
Point d'ébullition: 133…134 °C
Densité: 0,964
Point éclair: 33 °C (91 °F)
Numéro MDL: MFCD00005308
Numéro CAS: 20662-84-4
EINECS: 243-952-4
ADR: 3,III

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR