Imprimer
di-Sodium succinate hexahydraté ≥99%
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate
41983.30 41983.A1 41983.A7
41983.30EA 73.5 EUR
41983.30 41983.A1 41983.A7
di-Sodium succinate hexahydraté ≥99%
Succinate de disodium hexahydraté
Purity: 99% CAS No.: 6106-21-4 Molecular Formula: C4H4Na2O4.6H2O Formula Weight: 270.14 (162.05anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Melting Point: 120° -6H2O Beilstein Reference: 3921044 Merck Reference: 14,8679 MDL No.: MFCD00149117
Formule: NaOOCCH₂CH₂COONa·6H₂O
Poids moléculaire: 270.14 g/mol
Point de fusion: 120 °C (-6H₂O)
Densité: 1.529 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149117
Numéro CAS: 6106-21-4
EINECS: 205-778-7
Indice Merck: 13,08754