Imprimer

Tétrakis(triphénylphosphine)platine (0) (≥15,2% Pt)

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): Tetrakis(triphenylphosphine)platinum

10549.03 10549.06
10549.03EA 146 EUR
10549.03 10549.06
Tétrakis(triphénylphosphine)platine (0) (≥15,2% Pt)
Tétrakis(triphénylphosphine)platine (0)

CAS No.: 14221-02-4 Molecular Formula: Pt[P(C6H5)3]4 Formula Weight: 1244.21 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Sensitivity: Air Sensitive Appearance: Yellow Odour: Odourless Solubility: Soluble in benzene MDL No.: MFCD00010014


Hydrosilation and oxidation

Formule: C₇₂H₆₀P₄Pt
Numéro MDL: MFCD00010014
Numéro CAS: 14221-02-4
EINECS: 238-087-4

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR