Imprimer
Micro Curettes
Fournisseur: FINE SCIENCE TOOLS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10083-20 10082-15 10081-10 10080-05
FSCI10083-20EA 226 EUR
FSCI10083-20 FSCI10082-15 FSCI10081-10 FSCI10080-05
Micro Curettes
Curettes
Stainless steel. A curette tip consists of a scoop with sharp edges and is used for scraping or cleaning. It also can be used for separating and transporting, inoculating plates or transferring small amounts of chemicals.