Imprimer
All Weather Permanent Adhesive Vinyl Label Tape and Ribbon Kit for M6 Printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M6C1000595WT M6C2000595WT
BRDYM6C1000595WTEA 90.6 EUR
BRDYM6C1000595WT BRDYM6C2000595WT
All Weather Permanent Adhesive Vinyl Label Tape and Ribbon Kit for M6 Printers
Étiquettes
Jumpstart your wire and cable identification process with this material multipack that contains ribbons and rolls of indoor outdoor vinyl labels (B-595).
  • B-595 material is a durable, low-shrink vinyl with ultra-aggressive adhesive
  • For use on rough surfaces and in harsh environments
  • Material has an average outdoor durability of 8 to 10 years
  • Glossy finish is resistant to chemicals, grease, oil and cleaning agents
  • Ribbons can be changed in seconds and produce printed text that can withstand industrial conditions
Material Multipack includes: (3) rolls of M6C-500-595-WT, (2) M61-R10010 black ribbons. Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
Abrasion-resistant, chemical-resistant, oil-resistant, temperature range –4 to 82 ℃.
Applications: Electrical labeling, facility identification, floor marking, pipe and valve marking, warehouse marking.
Certifications: Compliant with Directive 2011/65/EU and 2015/863.