Imprimer
Wired Shelves, Norm 5
Fournisseur: HUPFER METALLWERKE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0101852 0101702 0101712 0101822
HUPF0101852EA 383 EUR
HUPF0101852 HUPF0101702 HUPF0101712 HUPF0101822
Wired Shelves, Norm 5
Mobilier Étagères
Stainless steel wire shelf, electropolished, consisting of 35×10×3,0 mm (1,4×0,4×0,12'') angle sections on the long side. Welded in between are Ø 3,5 mm (0,14'') wires running crosswise at distances of 25 mm (1,0'') with reinforcement struts on the underside made of Ø 3,5 mm ( 0,14'') wires.
  • Well ventilated storage
  • Perfect hygiene
  • High weight capacity
  • Easy handling