Imprimer
Universal Mini Circuit Breaker Lockouts
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
149515 149514
BRDY149515EA 98.1 EUR
BRDY149515 BRDY149514
Universal Mini Circuit Breaker Lockouts
Cadenas de sécurité
The Universal Mini Circuit Breaker Lockout is easily installed, requiring no tools and the low profile design allows panel door closure.
  • Material: Nylon
  • Lockout type: Tie-bar
  • Risk: Electrical risk
Available for single and multi-pole breakers. Attach a Brady safety padlock to complete lockout.