Imprimer
Winlog.Web Server Based Monitoring Software for EBI 25
Fournisseur: EBRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1340-2490 1340-2491
EBRO1340-2490EA 1420 EUR
EBRO1340-2490 EBRO1340-2491
Winlog.Web Server Based Monitoring Software for EBI 25
Logiciels Logiciels de laboratoire
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. Supports: data loggers EBI 25 series, MODBUS IP devices. Professional version for multiple users.
  • Connection of the interface IF 400 via USB and ethernet
  • FDA 21 CFR Part 11 functionality
  • Administration of large quantities of data
  • IQ-/OQ documentation available
  • Current version: 3.11
Web-based server solution: the measurement data can be read on all computers and smartphones which are connected with the network. Very flexible and comprehensive alarm management: alarm notifications due to specified events possible: alarm notification via email, visual and acoustical alarm via the user interface.
Hardware requirements processor: Pentium 2 GHz, main memory: 4 GB, free hard disk space: 20 GB, USB.
Software requirements: Windows 8,10 (32 bit or 64 bit).