Imprimer
Sterile Goods Baskets for ISO Units
Fournisseur: HUPFER METALLWERKE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7214006 7214016
HUPF7214006EA 335 EUR
HUPF7214006 HUPF7214016
Sterile Goods Baskets for ISO Units
Paniers Paniers en maille d'acier
Sterile supplies basket with insert frame for transporting, storing, and presenting sterile supplies in soft packaging without the use of containers.
  • Produced from electro-polished stainless steel
  • With U-shape profile rail compatible drawer frame
  • Stackable inside and on top of each other
Basket suitable for washing and sterilising devices, made from premium electro-polished round stainless steel material. Basket with upper insert frame suitable for sliding into L or U profile rails of cabinet systems, Norm 5 Medidul shelves or Hupfer Känguruh System®. The special design of the faces means the baskets can be stacked either one inside the other (when empty, for example during washing and disinfecting), or on top of each other (when filled).