Imprimer
Phalloidin lysine
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
5304 5305
AATB5304EA 137 EUR
AATB5304 AATB5305
Phalloidin lysine
Phalloidin lysine
Phalloidin lysine is a convenient building block that can be used for developing a variety of phalloidin derivatives that may be for studying cellular structures and other biological applications.
  • Useful in live or fixed cells
  • High affinity for F-actin
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab0bebd3f41402dd4e7d4a903da71d059ac500fc&vid=ac0dba370ad6322cb7a7df3bf3ccfc4b335dc018&prid=web&clid=SAPDAM