Imprimer
5-TAMRA alkyne ≥95%
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
487 961
AATB487EA 230 EUR
AATB487 AATB961
5-TAMRA alkyne ≥95%
5-TAMRA alkyne
5-TAMRA alkyne is an excellent azide-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Attention: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b4192865079abcc85874a09f3586600d1cec1f&vid=7444217d08247ac8bc7ec72a5447bdee3e3752d6&prid=web&clid=SAPDAM