Imprimer
California Red™ SE ≥90% Superior Replacement to Texas Red®
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
473 479
AATB473EA 230 EUR
AATB473 AATB479
California Red™ SE ≥90% Superior Replacement to Texas Red®
California Red™ SE
Although Texas Red® is the most popular labeling reagent of sulfonyl chloride, it is quite unstable in water.
  • Identical spectra to Texas Red
  • High Stability
  • Improved Labeling Efficiency
Attention: For research use only.