D-(-)-Luciferin potassium salt ≥95%
Marques : AAT BIOQUESTTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Luciferin is the most popular and versatile bioluminescent substrate.
Formule:
C₁₁H₇KN₂O₃S₂ |
Numéro MDL:
MFCD00044928 Numéro CAS: 115144-35-9 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...