Imprimer

4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate

Marques : AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): Disodium 4-methylumbelliferyl phosphate trihydrate, 4-MUP trihydrate

11610 11612
AATB11610EA 120 EUR
AATB11610 AATB11612
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate

A fluorogenic substrate for phosphatases.

Formule: C₁₀H₇Na₂O₆P·3H₂O
Numéro MDL: MFCD00037614
Numéro CAS: 22919-26-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3060efa2c21773bda89a4888d06cec757ef65afc&vid=558a0ef4f7cdf9e2e629cc389b7d6b7b88d2a540&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR