Imprimer
iFluor® 594, Succinimidyl Ester
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
71029 1029 71508 71558
AATB71029EA 114 EUR
AATB71029 AATB1029 AATB71508 AATB71558
iFluor® 594, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Attention: For research use only.